Product Details

SNP ID
rs148160279
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:46274049 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGGGAGCAGCTGCACGTGTCTGC[A/G]GTAGAGACATTTGCCAATGGGTGAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TTC38 PubMed Links

Gene Details

Gene
TTC38
Gene Name
tetratricopeptide repeat domain 38
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017931.2 421 Silent Mutation GCA,GCG A115A NP_060401.2
XM_011530259.1 421 Silent Mutation GCA,GCG A50A XP_011528561.1
XM_011530260.2 421 Silent Mutation GCA,GCG A115A XP_011528562.1

View Full Product Details