Product Details

SNP ID
rs148337668
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:38485969 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGGAGGAGGAGGGGGAGGAGGA[A/G]GAGGGTATTGGTAGGCAGTCTGCCC
Phenotype
MIM: 608469
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DDX17 PubMed Links

Gene Details

Gene
DDX17
Gene Name
DEAD-box helicase 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098504.1 2265 Missense Mutation CCT,CTT P721L NP_001091974.1
NM_006386.4 2265 Missense Mutation CCT,CTT P719L NP_006377.2
Gene
KDELR3
Gene Name
KDEL endoplasmic reticulum protein retention receptor 3
There are no transcripts associated with this gene.

View Full Product Details