Product Details

SNP ID
rs148781703
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:37181821 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATAGACTGACCCCATCAGCATCA[A/C]CCAGGAACATATGAGAAACACAGAA
Phenotype
MIM: 614910
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C1QTNF6 PubMed Links

Gene Details

Gene
C1QTNF6
Gene Name
C1q and tumor necrosis factor related protein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031910.3 3775 UTR 3 NP_114116.3
NM_182486.1 3775 Intron NP_872292.1
XM_011529857.1 3775 UTR 3 XP_011528159.1
XM_017028569.1 3775 UTR 3 XP_016884058.1
XM_017028570.1 3775 UTR 3 XP_016884059.1
XM_017028571.1 3775 UTR 3 XP_016884060.1
XM_017028572.1 3775 UTR 3 XP_016884061.1
XM_017028573.1 3775 UTR 3 XP_016884062.1
XM_017028574.1 3775 UTR 3 XP_016884063.1

View Full Product Details