Product Details
- SNP ID
-
rs149777993
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:45285431 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAATATTGGGCTGTCCTGGGAACTG[C/T]CAGTCCATGGGACCAGCTGGAGCCT
- Phenotype
-
MIM: 611559
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
UPK3A
PubMed Links
Gene Details
- Gene
- UPK3A
- Gene Name
- uroplakin 3A
View Full Product Details