Product Details

SNP ID
rs149988472
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.22:42516072 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCTTTGGGCTCTCAGCCAGGTCC[A/G]GCTTCTCCTGCAACTCTACAGACTG
Phenotype
MIM: 607979
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
RRP7A PubMed Links
Additional Information
For this assay, SNP(s) [rs11553441,rs1812240] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
RRP7A
Gene Name
ribosomal RNA processing 7 homolog A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015703.4 318 Missense Mutation CCG,CTG P94L NP_056518.2
Gene
SERHL
Gene Name
serine hydrolase-like (pseudogene)
There are no transcripts associated with this gene.

View Full Product Details