Product Details

SNP ID
rs150137904
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:50217804 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTCCTCCAGGTGGGGCTGGTAGC[C/T]GCGGTTCACCAGCTTGGTCACCACT
Phenotype
MIM: 607917 MIM: 610053
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SELO PubMed Links

Gene Details

Gene
SELO
Gene Name
selenoprotein O
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031454.1 5904 Intron NP_113642.1
Gene
TUBGCP6
Gene Name
tubulin gamma complex associated protein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020461.3 5904 Missense Mutation AGC,GGC S1798G NP_065194.2

View Full Product Details