Product Details

SNP ID
rs150320937
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:19764275 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCGCAGTGGATGAAGCAAATCGT[A/G]TCCTTCGACAAGCTCAAGCTGACCA
Phenotype
MIM: 602054
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TBX1 PubMed Links

Gene Details

Gene
TBX1
Gene Name
T-box 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005992.1 897 Silent Mutation GTA,GTG V211V NP_005983.1
NM_080646.1 897 Silent Mutation GTA,GTG V211V NP_542377.1
NM_080647.1 897 Silent Mutation GTA,GTG V211V NP_542378.1
XM_006724312.2 897 Silent Mutation GTA,GTG V211V XP_006724375.1
XM_017028925.1 897 Silent Mutation GTA,GTG V261V XP_016884414.1
XM_017028926.1 897 Silent Mutation GTA,GTG V211V XP_016884415.1
XM_017028927.1 897 Intron XP_016884416.1
XM_017028928.1 897 Silent Mutation GTA,GTG V261V XP_016884417.1

View Full Product Details