Product Details

SNP ID
rs150532024
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:35739469 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGCAGAATATAACACAAGAAAAA[C/G]AATTCATATCCCTTAGGGAAAAAAG
Phenotype
MIM: 607255 MIM: 612149
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
APOL5 PubMed Links

Gene Details

Gene
APOL5
Gene Name
apolipoprotein L5
There are no transcripts associated with this gene.

Gene
RBFOX2
Gene Name
RNA binding protein, fox-1 homolog 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031695.2 6268 UTR 3 NP_001026865.1
NM_001082576.1 6268 UTR 3 NP_001076045.1
NM_001082577.1 6268 UTR 3 NP_001076046.1
NM_001082578.1 6268 UTR 3 NP_001076047.1
NM_001082579.1 6268 UTR 3 NP_001076048.1
NM_014309.2 6268 UTR 3 NP_055124.1
XM_005261428.2 6268 UTR 3 XP_005261485.1
XM_005261429.2 6268 UTR 3 XP_005261486.1
XM_005261430.2 6268 UTR 3 XP_005261487.1
XM_005261431.2 6268 UTR 3 XP_005261488.1
XM_005261432.2 6268 UTR 3 XP_005261489.1
XM_005261433.2 6268 UTR 3 XP_005261490.1
XM_005261435.1 6268 UTR 3 XP_005261492.1
XM_005261437.1 6268 UTR 3 XP_005261494.1
XM_006724185.2 6268 UTR 3 XP_006724248.1
XM_006724186.2 6268 UTR 3 XP_006724249.1
XM_006724187.2 6268 UTR 3 XP_006724250.1
XM_006724188.2 6268 UTR 3 XP_006724251.1
XM_006724189.3 6268 UTR 3 XP_006724252.1
XM_006724190.1 6268 UTR 3 XP_006724253.1
XM_006724191.1 6268 UTR 3 XP_006724254.1
XM_006724192.2 6268 UTR 3 XP_006724255.1
XM_006724193.3 6268 Intron XP_006724256.1
XM_006724194.2 6268 UTR 3 XP_006724257.1
XM_017028686.1 6268 UTR 3 XP_016884175.1
XM_017028687.1 6268 UTR 3 XP_016884176.1
XM_017028688.1 6268 UTR 3 XP_016884177.1
XM_017028689.1 6268 UTR 3 XP_016884178.1
XM_017028690.1 6268 UTR 3 XP_016884179.1
XM_017028691.1 6268 UTR 3 XP_016884180.1
XM_017028692.1 6268 UTR 3 XP_016884181.1
XM_017028693.1 6268 UTR 3 XP_016884182.1
XM_017028694.1 6268 UTR 3 XP_016884183.1
XM_017028695.1 6268 UTR 3 XP_016884184.1
XM_017028696.1 6268 UTR 3 XP_016884185.1
XM_017028697.1 6268 UTR 3 XP_016884186.1
XM_017028698.1 6268 UTR 3 XP_016884187.1

View Full Product Details