Product Details
- SNP ID
-
rs137894462
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:218357703 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCATGCCTCTAGCCGAGGCTTCTT[A/G]GACAAAATGCTCTGCGGAAATTCCC
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CATIP
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs145097529] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CATIP
- Gene Name
- ciliogenesis associated TTC17 interacting protein
- Gene
- CATIP-AS1
- Gene Name
- CATIP antisense RNA 1
There are no transcripts associated with this gene.
- Gene
- CATIP-AS2
- Gene Name
- CATIP antisense RNA 2
There are no transcripts associated with this gene.
View Full Product Details