Product Details

SNP ID
rs138220608
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:196809774 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAAAAGAAATTACTTCAGATACC[A/G]AGCTGAGAAATACACCTTATACACA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C2orf66 PubMed Links

Gene Details

Gene
C2orf66
Gene Name
chromosome 2 open reading frame 66
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_213608.2 503 UTR 5 NP_998773.1

View Full Product Details