Product Details

SNP ID
rs139256632
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:231047396 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGGGCACCCATCGTTGAGAGTGC[A/T]GCTGGGAAGAACTCTGAACCAGAAG
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
C2orf72 PubMed Links

Gene Details

Gene
C2orf72
Gene Name
chromosome 2 open reading frame 72
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144994.1 1263 UTR 3 NP_001138466.1
Gene
PSMD1
Gene Name
proteasome 26S subunit, non-ATPase 1
There are no transcripts associated with this gene.

View Full Product Details