Product Details

SNP ID
rs139268416
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:30231757 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACTTCATGTCTATATATTTCCCC[A/T]TTCACTGGTGAGTACCCTGCGCCTG
Phenotype
MIM: 611763
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LBH PubMed Links

Gene Details

Gene
LBH
Gene Name
limb bud and heart development
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030915.3 227 Missense Mutation ATT,TTT I7F NP_112177.2
XM_017005059.1 227 Intron XP_016860548.1

View Full Product Details