Product Details

SNP ID
rs139624409
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:232480429 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCTCAGGGGCATGCTTGTCAGTC[A/G]GCACCTGCAGGTAGATGGACTGCGA
Phenotype
MIM: 605896
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ECEL1 PubMed Links

Gene Details

Gene
ECEL1
Gene Name
endothelin converting enzyme like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290787.1 2434 Missense Mutation CCG,CTG P731L NP_001277716.1
NM_004826.3 2434 Missense Mutation CCG,CTG P733L NP_004817.2

View Full Product Details