Product Details

SNP ID
rs139755676
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27132076 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTGCCCAGGCCTAGGAAGGTGCC[A/G]GATTCACTGCAACAAACAATAAAGA
Phenotype
MIM: 612195 MIM: 606395
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABHD1 PubMed Links

Gene Details

Gene
ABHD1
Gene Name
abhydrolase domain containing 1
There are no transcripts associated with this gene.

Gene
PREB
Gene Name
prolactin regulatory element binding
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_006711914.2 993 Silent Mutation TCC,TCT S253S XP_006711977.1
XM_011532471.2 993 Silent Mutation TCC,TCT S311S XP_011530773.1
XM_011532472.2 993 Silent Mutation TCC,TCT S253S XP_011530774.1
Gene
PRR30
Gene Name
proline rich 30
There are no transcripts associated with this gene.

View Full Product Details