Product Details

SNP ID
rs139821106
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:211376871 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAGCCACGCAATCCCTGGTGCTC[G/T]CAACATGAGAAGGAGACACACCAGC
Phenotype
MIM: 600543
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ERBB4 PubMed Links

Gene Details

Gene
ERBB4
Gene Name
erb-b2 receptor tyrosine kinase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042599.1 11318 UTR 3 NP_001036064.1
NM_005235.2 11318 UTR 3 NP_005226.1
XM_005246376.2 11318 UTR 3 XP_005246433.1
XM_005246377.2 11318 UTR 3 XP_005246434.1
XM_006712364.2 11318 UTR 3 XP_006712427.1
XM_017003577.1 11318 UTR 3 XP_016859066.1
XM_017003578.1 11318 UTR 3 XP_016859067.1
XM_017003579.1 11318 UTR 3 XP_016859068.1
XM_017003580.1 11318 UTR 3 XP_016859069.1
XM_017003581.1 11318 UTR 3 XP_016859070.1
XM_017003582.1 11318 UTR 3 XP_016859071.1

View Full Product Details