Product Details

SNP ID
rs140167417
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:71133395 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACAAAGGACAGGGAAAACCAAGAG[A/T]AAAGTCCATAGTAGATGATAAATTC
Phenotype
MIM: 608419 MIM: 605503
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MCEE PubMed Links

Gene Details

Gene
MCEE
Gene Name
methylmalonyl-CoA epimerase
There are no transcripts associated with this gene.

Gene
MPHOSPH10
Gene Name
M-phase phosphoprotein 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005791.2 939 Missense Mutation GAA,GTA E196V NP_005782.1

View Full Product Details