Product Details

SNP ID
rs140404341
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:218871197 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCAGTTCCAGTTCCGCTTCCGCC[A/G]CTGGAATTGCTCCAGCCACAGCAAG
Phenotype
MIM: 606268 MIM: 604663
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
WNT10A PubMed Links

Gene Details

Gene
WNT10A
Gene Name
Wnt family member 10A
There are no transcripts associated with this gene.

Gene
WNT6
Gene Name
Wnt family member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006522.3 466 Missense Mutation CAC,CGC H84R NP_006513.1

View Full Product Details