Product Details

SNP ID
rs140500963
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:57912830 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAATATTTGTATCCCGAGATTCAT[A/C]TATTAAAACCCTTACCCCTAATGTG
Phenotype
MIM: 602169
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
VRK2 PubMed Links

Gene Details

Gene
VRK2
Gene Name
vaccinia related kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130480.2 Intron NP_001123952.1
NM_001130481.2 Intron NP_001123953.1
NM_001130482.2 Intron NP_001123954.1
NM_001130483.2 Intron NP_001123955.1
NM_001288836.1 Intron NP_001275765.1
NM_001288837.1 Intron NP_001275766.1
NM_001288838.1 Intron NP_001275767.1
NM_001288839.1 Intron NP_001275768.1
NM_006296.6 Intron NP_006287.2
XM_005264540.4 Intron XP_005264597.1
XM_006712090.3 Intron XP_006712153.1
XM_006712091.3 Intron XP_006712154.1
XM_006712092.3 Intron XP_006712155.1
XM_006712093.3 Intron XP_006712156.1
XM_011533092.2 Intron XP_011531394.1
XM_017004858.1 Intron XP_016860347.1
XM_017004859.1 Intron XP_016860348.1
XM_017004860.1 Intron XP_016860349.1
XM_017004861.1 Intron XP_016860350.1
XM_017004862.1 Intron XP_016860351.1
XM_017004863.1 Intron XP_016860352.1
XM_017004864.1 Intron XP_016860353.1
XM_017004865.1 Intron XP_016860354.1
XM_017004866.1 Intron XP_016860355.1

View Full Product Details