Product Details

SNP ID
rs140925410
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:207724857 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAGGGCAAGCACAATTTTCCTGA[A/G]CAAATCTCAAACGGATGGTAAGACA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCNYL1 PubMed Links

Gene Details

Gene
CCNYL1
Gene Name
cyclin Y like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142300.1 383 Missense Mutation AAC,AGC N93S NP_001135772.1
NM_152523.2 383 Missense Mutation AAC,AGC N23S NP_689736.1
XM_005246343.1 383 Intron XP_005246400.1

View Full Product Details