Product Details

SNP ID
rs141107432
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:218675471 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGTGGCATCAAGCTCTGTGACTT[C/T]GGGTAAAGATTCTGAGCATCCATCT
Phenotype
MIM: 616014 MIM: 607652
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RNF25 PubMed Links

Gene Details

Gene
RNF25
Gene Name
ring finger protein 25
There are no transcripts associated with this gene.

Gene
STK36
Gene Name
serine/threonine kinase 36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243313.1 379 Silent Mutation TTC,TTT F144F NP_001230242.1
NM_015690.4 379 Silent Mutation TTC,TTT F144F NP_056505.2
XM_005246464.1 379 Silent Mutation TTC,TTT F144F XP_005246521.1
XM_011510959.1 379 Silent Mutation TTC,TTT F144F XP_011509261.1
XM_011510960.1 379 Silent Mutation TTC,TTT F90F XP_011509262.1
XM_011510961.1 379 Intron XP_011509263.1
XM_017003804.1 379 Silent Mutation TTC,TTT F144F XP_016859293.1
XM_017003805.1 379 UTR 5 XP_016859294.1

View Full Product Details