Product Details

SNP ID
rs142143856
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:218358251 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGCAATGTAAAGCTCAAAAGCAA[C/T]GCACTTCAGAAAACTGATTTTAAAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CATIP PubMed Links

Gene Details

Gene
CATIP
Gene Name
ciliogenesis associated TTC17 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320865.1 Intron NP_001307794.1
NM_198559.1 Intron NP_940961.1
XM_005246539.4 Intron XP_005246596.1
XM_005246541.4 Intron XP_005246598.1
XM_011511148.2 Intron XP_011509450.1
XM_011511149.2 Intron XP_011509451.1
XM_011511150.2 Intron XP_011509452.1
XM_017004053.1 Intron XP_016859542.1
Gene
CATIP-AS1
Gene Name
CATIP antisense RNA 1
There are no transcripts associated with this gene.

Gene
CATIP-AS2
Gene Name
CATIP antisense RNA 2
There are no transcripts associated with this gene.

View Full Product Details