Product Details
- SNP ID
-
rs142150744
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:229768717 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATCTCAAACAAAAACTTCACAGCC[C/T]GACTATAACAGAAATAAATATACCA
- Phenotype
-
MIM: 604506
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TRIP12
PubMed Links
Gene Details
- Gene
- TRIP12
- Gene Name
- thyroid hormone receptor interactor 12
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001284214.1 |
5949 |
Missense Mutation |
CAG,CGG |
Q1942R |
NP_001271143.1 |
NM_001284215.1 |
5949 |
Missense Mutation |
CAG,CGG |
Q1927R |
NP_001271144.1 |
NM_001284216.1 |
5949 |
Missense Mutation |
CAG,CGG |
Q1624R |
NP_001271145.1 |
NM_004238.2 |
5949 |
Missense Mutation |
CAG,CGG |
Q1894R |
NP_004229.1 |
XM_005246954.4 |
5949 |
Missense Mutation |
CAG,CGG |
Q1970R |
XP_005247011.1 |
XM_005246955.4 |
5949 |
Missense Mutation |
CAG,CGG |
Q1970R |
XP_005247012.1 |
XM_005246956.4 |
5949 |
Missense Mutation |
CAG,CGG |
Q1969R |
XP_005247013.1 |
XM_005246957.4 |
5949 |
Missense Mutation |
CAG,CGG |
Q1965R |
XP_005247014.1 |
XM_005246958.4 |
5949 |
Missense Mutation |
CAG,CGG |
Q1943R |
XP_005247015.1 |
XM_005246960.4 |
5949 |
Missense Mutation |
CAG,CGG |
Q1941R |
XP_005247017.1 |
XM_005246961.3 |
5949 |
Missense Mutation |
CAG,CGG |
Q1928R |
XP_005247018.1 |
XM_005246962.4 |
5949 |
Missense Mutation |
CAG,CGG |
Q1927R |
XP_005247019.1 |
XM_006712852.3 |
5949 |
Missense Mutation |
CAG,CGG |
Q1928R |
XP_006712915.1 |
XM_011512180.2 |
5949 |
Missense Mutation |
CAG,CGG |
Q1970R |
XP_011510482.1 |
XM_011512181.2 |
5949 |
Missense Mutation |
CAG,CGG |
Q1970R |
XP_011510483.1 |
XM_011512183.2 |
5949 |
Missense Mutation |
CAG,CGG |
Q1928R |
XP_011510485.1 |
XM_017005272.1 |
5949 |
Missense Mutation |
CAG,CGG |
Q1970R |
XP_016860761.1 |
XM_017005273.1 |
5949 |
Missense Mutation |
CAG,CGG |
Q1970R |
XP_016860762.1 |
XM_017005274.1 |
5949 |
Missense Mutation |
CAG,CGG |
Q1970R |
XP_016860763.1 |
XM_017005275.1 |
5949 |
Missense Mutation |
CAG,CGG |
Q1969R |
XP_016860764.1 |
XM_017005276.1 |
5949 |
Missense Mutation |
CAG,CGG |
Q1969R |
XP_016860765.1 |
XM_017005277.1 |
5949 |
Missense Mutation |
CAG,CGG |
Q1969R |
XP_016860766.1 |
XM_017005278.1 |
5949 |
Missense Mutation |
CAG,CGG |
Q1969R |
XP_016860767.1 |
XM_017005279.1 |
5949 |
Missense Mutation |
CAG,CGG |
Q1964R |
XP_016860768.1 |
XM_017005280.1 |
5949 |
Missense Mutation |
CAG,CGG |
Q1964R |
XP_016860769.1 |
XM_017005281.1 |
5949 |
Missense Mutation |
CAG,CGG |
Q1942R |
XP_016860770.1 |
XM_017005282.1 |
5949 |
Missense Mutation |
CAG,CGG |
Q1941R |
XP_016860771.1 |
XM_017005283.1 |
5949 |
Missense Mutation |
CAG,CGG |
Q1937R |
XP_016860772.1 |
XM_017005284.1 |
5949 |
Missense Mutation |
CAG,CGG |
Q1935R |
XP_016860773.1 |
XM_017005285.1 |
5949 |
Missense Mutation |
CAG,CGG |
Q1928R |
XP_016860774.1 |
XM_017005286.1 |
5949 |
Missense Mutation |
CAG,CGG |
Q1927R |
XP_016860775.1 |
XM_017005287.1 |
5949 |
Missense Mutation |
CAG,CGG |
Q1922R |
XP_016860776.1 |
XM_017005288.1 |
5949 |
Missense Mutation |
CAG,CGG |
Q1922R |
XP_016860777.1 |
XM_017005289.1 |
5949 |
Missense Mutation |
CAG,CGG |
Q1914R |
XP_016860778.1 |
XM_017005290.1 |
5949 |
Missense Mutation |
CAG,CGG |
Q1913R |
XP_016860779.1 |
XM_017005291.1 |
5949 |
Missense Mutation |
CAG,CGG |
Q1899R |
XP_016860780.1 |
XM_017005292.1 |
5949 |
Missense Mutation |
CAG,CGG |
Q1895R |
XP_016860781.1 |
XM_017005293.1 |
5949 |
Missense Mutation |
CAG,CGG |
Q1900R |
XP_016860782.1 |
View Full Product Details