Product Details

SNP ID
rs142161992
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:17670536 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTTCACCTCCAGACAAGGCTCTCA[C/T]GTCATTGAAAGCAGCTTTATTTCCT
Phenotype
MIM: 609387
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SMC6 PubMed Links

Gene Details

Gene
SMC6
Gene Name
structural maintenance of chromosomes 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142286.1 3074 Missense Mutation NP_001135758.1
NM_024624.5 3074 Missense Mutation NP_078900.1
XM_011533107.2 3074 Missense Mutation XP_011531409.1
XM_011533108.2 3074 Missense Mutation XP_011531410.1
XM_017004913.1 3074 Missense Mutation XP_016860402.1
XM_017004914.1 3074 Missense Mutation XP_016860403.1
XM_017004915.1 3074 Missense Mutation XP_016860404.1
XM_017004916.1 3074 Missense Mutation XP_016860405.1

View Full Product Details