Product Details

SNP ID
rs142170310
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27082158 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGACCAAGGAGCTGCAAGGCCTGC[G/T]GGGCGTCCTGCAAGGACTGAGCGGG
Phenotype
MIM: 130660 MIM: 614058
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
EMILIN1 PubMed Links

Gene Details

Gene
EMILIN1
Gene Name
elastin microfibril interfacer 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007046.3 1086 Missense Mutation CGG,CTG R196L NP_008977.1
Gene
KHK
Gene Name
ketohexokinase
There are no transcripts associated with this gene.

View Full Product Details