Product Details

SNP ID
rs142321560
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:158977816 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATCTGGACCCTAACTTGTTTGCA[A/G]GCCCAGCTGTCCTCTCCTTGTCTCT
Phenotype
MIM: 611397
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TANC1 PubMed Links

Gene Details

Gene
TANC1
Gene Name
tetratricopeptide repeat, ankyrin repeat and coiled-coil containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145909.1 Intron NP_001139381.1
NM_033394.2 Intron NP_203752.2
XM_005246918.4 Intron XP_005246975.1
XM_006712810.3 Intron XP_006712873.1
XM_006712812.3 Intron XP_006712875.1
XM_006712814.3 Intron XP_006712877.1
XM_011512048.2 Intron XP_011510350.1
XM_011512051.2 Intron XP_011510353.1
XM_011512053.2 Intron XP_011510355.1
XM_011512055.2 Intron XP_011510357.1
XM_011512058.2 Intron XP_011510360.1
XM_011512059.2 Intron XP_011510361.1
XM_011512062.2 Intron XP_011510364.1
XM_017005141.1 Intron XP_016860630.1
XM_017005142.1 Intron XP_016860631.1
XM_017005143.1 Intron XP_016860632.1
XM_017005144.1 Intron XP_016860633.1
XM_017005145.1 Intron XP_016860634.1
XM_017005146.1 Intron XP_016860635.1
XM_017005147.1 Intron XP_016860636.1
XM_017005148.1 Intron XP_016860637.1
XM_017005149.1 Intron XP_016860638.1
XM_017005150.1 Intron XP_016860639.1
XM_017005151.1 Intron XP_016860640.1
XM_017005152.1 Intron XP_016860641.1
XM_017005153.1 Intron XP_016860642.1
XM_017005154.1 Intron XP_016860643.1
XM_017005155.1 Intron XP_016860644.1
XM_017005156.1 Intron XP_016860645.1
XM_017005157.1 Intron XP_016860646.1
XM_017005158.1 Intron XP_016860647.1

View Full Product Details