Product Details

SNP ID
rs142457164
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:9845286 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGGGTCTTACTGATGAAGGCAAA[C/T]ATTATTGCACTTCTTGCCACAATGT
Phenotype
MIM: 604904
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TAF1B PubMed Links

Gene Details

Gene
TAF1B
Gene Name
TATA-box binding protein associated factor, RNA polymerase I subunit B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318976.1 104 UTR 5 NP_001305905.1
NM_001318977.1 104 UTR 5 NP_001305906.1
NM_005680.2 104 Missense Mutation CAT,TAT H29Y NP_005671.2
XM_011510410.2 104 Missense Mutation CAT,TAT H29Y XP_011508712.1
XM_011510411.1 104 UTR 5 XP_011508713.1

View Full Product Details