Product Details
- SNP ID
-
rs142510036
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:24819977 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTGGGGCAGTGTGACAGAGGGGCC[A/G]TTGGGGAAGGTGGCTAGCTTATCCC
- Phenotype
-
MIM: 600291
MIM: 611504
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ADCY3
PubMed Links
Gene Details
- Gene
- ADCY3
- Gene Name
- adenylate cyclase 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001320613.1 |
2036 |
Silent Mutation |
AAC,AAT |
N1131N |
NP_001307542.1 |
NM_004036.4 |
2036 |
Silent Mutation |
AAC,AAT |
N1130N |
NP_004027.2 |
XM_006711925.1 |
2036 |
Silent Mutation |
AAC,AAT |
N1153N |
XP_006711988.1 |
XM_011532489.1 |
2036 |
Silent Mutation |
AAC,AAT |
N1172N |
XP_011530791.1 |
XM_011532490.1 |
2036 |
Silent Mutation |
AAC,AAT |
N1171N |
XP_011530792.1 |
XM_011532491.1 |
2036 |
Silent Mutation |
AAC,AAT |
N1150N |
XP_011530793.1 |
XM_011532492.2 |
2036 |
UTR 3 |
|
|
XP_011530794.1 |
XM_011532493.1 |
2036 |
Silent Mutation |
AAC,AAT |
N1126N |
XP_011530795.1 |
XM_011532494.1 |
2036 |
Silent Mutation |
AAC,AAT |
N1106N |
XP_011530796.1 |
XM_011532496.1 |
2036 |
Silent Mutation |
AAC,AAT |
N931N |
XP_011530798.1 |
XM_017003186.1 |
2036 |
Silent Mutation |
AAC,AAT |
N1152N |
XP_016858675.1 |
XM_017003187.1 |
2036 |
Silent Mutation |
AAC,AAT |
N1149N |
XP_016858676.1 |
XM_017003188.1 |
2036 |
UTR 3 |
|
|
XP_016858677.1 |
XM_017003189.1 |
2036 |
Silent Mutation |
AAC,AAT |
N1125N |
XP_016858678.1 |
XM_017003190.1 |
2036 |
Silent Mutation |
AAC,AAT |
N1084N |
XP_016858679.1 |
XM_017003191.1 |
2036 |
Silent Mutation |
AAC,AAT |
N960N |
XP_016858680.1 |
XM_017003192.1 |
2036 |
Silent Mutation |
AAC,AAT |
N890N |
XP_016858681.1 |
XM_017003193.1 |
2036 |
Silent Mutation |
AAC,AAT |
N889N |
XP_016858682.1 |
- Gene
- CENPO
- Gene Name
- centromere protein O
View Full Product Details