Product Details

SNP ID
rs142589700
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:18555440 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGTGGAATGTGTATGTGCCTCCC[C/T]AGATTTGTCCGTACAATACCAGGAT
Phenotype
MIM: 610526 MIM: 616796
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NT5C1B PubMed Links

Gene Details

Gene
NT5C1B
Gene Name
5'-nucleotidase, cytosolic IB
There are no transcripts associated with this gene.

Gene
NT5C1B-RDH14
Gene Name
NT5C1B-RDH14 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199103.1 883 Silent Mutation CTA,CTG L568L NP_001186032.1
NM_001199104.1 883 UTR 3 NP_001186033.1
Gene
RDH14
Gene Name
retinol dehydrogenase 14 (all-trans/9-cis/11-cis)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020905.3 883 Silent Mutation CTA,CTG L254L NP_065956.1

View Full Product Details