Product Details
- SNP ID
-
rs142787154
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:68158414 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAGAAACAAGGAAAATTCCAGTCC[C/T]AGCTAACAGATACACACCATTGAAA
- Phenotype
-
MIM: 610729
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PNO1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs7590838] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PNO1
- Gene Name
- partner of NOB1 homolog
There are no transcripts associated with this gene.
- Gene
- WDR92
- Gene Name
- WD repeat domain 92
View Full Product Details