Product Details
- SNP ID
-
rs143173370
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:218275482 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAATGGGGGCTTGCTCCTTAATTG[C/T]GATCCCCCATCAGCTGCAGCACAAA
- Phenotype
-
MIM: 603488
MIM: 609023
MIM: 610364
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
AAMP
PubMed Links
Gene Details
- Gene
- AAMP
- Gene Name
- angio associated migratory cell protein
There are no transcripts associated with this gene.
- Gene
- MIR6513
- Gene Name
- microRNA 6513
There are no transcripts associated with this gene.
- Gene
- PNKD
- Gene Name
- paroxysmal nonkinesigenic dyskinesia
- Gene
- TMBIM1
- Gene Name
- transmembrane BAX inhibitor motif containing 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001321427.1 |
1006 |
Missense Mutation |
CAC,CGC |
H310R |
NP_001308356.1 |
NM_001321428.1 |
1006 |
Missense Mutation |
CAC,CGC |
H310R |
NP_001308357.1 |
NM_001321429.1 |
1006 |
Missense Mutation |
CAC,CGC |
H310R |
NP_001308358.1 |
NM_001321430.1 |
1006 |
Missense Mutation |
CAC,CGC |
H310R |
NP_001308359.1 |
NM_001321432.1 |
1006 |
Missense Mutation |
CAC,CGC |
H310R |
NP_001308361.1 |
NM_001321433.1 |
1006 |
Missense Mutation |
CAC,CGC |
H310R |
NP_001308362.1 |
NM_001321435.1 |
1006 |
Missense Mutation |
CAC,CGC |
H310R |
NP_001308364.1 |
NM_001321436.1 |
1006 |
Missense Mutation |
CAC,CGC |
H310R |
NP_001308365.1 |
NM_001321438.1 |
1006 |
Missense Mutation |
CAC,CGC |
H199R |
NP_001308367.1 |
NM_022152.5 |
1006 |
Missense Mutation |
CAC,CGC |
H310R |
NP_071435.2 |
XM_011511627.1 |
1006 |
Missense Mutation |
CAC,CGC |
H310R |
XP_011509929.1 |
View Full Product Details