Product Details
- SNP ID
-
rs143283662
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:85343319 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGTACAAGTTCCGCTTCAGGATGG[C/T]GCTGCTGCACAGCAGGGCACCTTGC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RETSAT
PubMed Links
Gene Details
- Gene
- RETSAT
- Gene Name
- retinol saturase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_017750.3 |
1947 |
Missense Mutation |
ACC,GCC |
T586A |
NP_060220.3 |
View Full Product Details