Product Details

SNP ID
rs143284239
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:127947570 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATAAATTTGGAATGTCCTTGCCT[A/G]TATTTTAAAAGACACTGCAGAGAAA
Phenotype
MIM: 609697
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
SAP130 PubMed Links

Gene Details

Gene
SAP130
Gene Name
Sin3A associated protein 130
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145928.1 Intron NP_001139400.1
NM_024545.3 Intron NP_078821.2
XM_005263767.3 Intron XP_005263824.1
XM_005263768.3 Intron XP_005263825.1
XM_005263769.3 Intron XP_005263826.1
XM_005263770.3 Intron XP_005263827.1
XM_006712748.3 Intron XP_006712811.1
XM_006712749.3 Intron XP_006712812.1
XM_017004903.1 Intron XP_016860392.1
XM_017004904.1 Intron XP_016860393.1
XM_017004905.1 Intron XP_016860394.1

View Full Product Details