Product Details

SNP ID
rs143590376
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:24183581 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCTACTTCACTTTCACTTCACAC[A/T]GTGTGATTCCAAAAGAGTGGCATAA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FAM228A PubMed Links

Gene Details

Gene
FAM228A
Gene Name
family with sequence similarity 228 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040710.2 483 Missense Mutation AGT,TGT S113C NP_001035800.1

View Full Product Details