Product Details

SNP ID
rs143667380
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:161416464 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTATCATGCTCTCCAAGAAATTTCT[C/G]AATGTGAGCAGCAGCTACCCACATT
Phenotype
MIM: 607173 MIM: 604616
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PSMD14 PubMed Links

Gene Details

Gene
PSMD14
Gene Name
proteasome 26S subunit, non-ATPase 14
There are no transcripts associated with this gene.

Gene
TBR1
Gene Name
T-box, brain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006593.3 371 Silent Mutation CTC,CTG L18L NP_006584.1

View Full Product Details