Product Details

SNP ID
rs144041023
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:127706366 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCCGTGAAGCTCCTCGCCTCGGC[A/G]GGCCAGAGTTCATGTTACTCCCTCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SFT2D3 PubMed Links

Gene Details

Gene
SFT2D3
Gene Name
SFT2 domain containing 3
There are no transcripts associated with this gene.

Gene
WDR33
Gene Name
WD repeat domain 33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001006622.2 4167 Intron NP_001006623.1
NM_001006623.2 4167 Intron NP_001006624.1
NM_018383.4 4167 Missense Mutation CCG,CTG P1323L NP_060853.3
XM_005263697.2 4167 UTR 3 XP_005263754.1
XM_011511436.1 4167 Missense Mutation CCG,CTG P1323L XP_011509738.1
XM_017004435.1 4167 Intron XP_016859924.1
XM_017004436.1 4167 Intron XP_016859925.1

View Full Product Details