Product Details

SNP ID
rs144172331
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:208142738 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCCCCGGCCTCAGCAGATACTGC[C/T]TCCCCCTGTAGTTGGGCATCTCATA
Phenotype
MIM: 123670 MIM: 123680
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CRYGB PubMed Links

Gene Details

Gene
CRYGB
Gene Name
crystallin gamma B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005210.3 461 Missense Mutation AAG,AGG K143R NP_005201.2
XM_017003402.1 461 Missense Mutation AAG,AGG K145R XP_016858891.1
Gene
CRYGC
Gene Name
crystallin gamma C
There are no transcripts associated with this gene.

Gene
LOC100507443
Gene Name
uncharacterized LOC100507443
There are no transcripts associated with this gene.

View Full Product Details