Product Details

SNP ID
rs144354033
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:165748266 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATTTGACAGACAAGTAAACCAAA[C/T]GCAAAAAAAAGTTCACCTGCATTTT
Phenotype
MIM: 601756
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
GALNT3 PubMed Links

Gene Details

Gene
GALNT3
Gene Name
polypeptide N-acetylgalactosaminyltransferase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004482.3 2687 UTR 3 NP_004473.2
XM_005246449.1 2687 UTR 3 XP_005246506.1
XM_011510929.1 2687 UTR 3 XP_011509231.1
XM_017003770.1 2687 UTR 3 XP_016859259.1

View Full Product Details