Product Details
- SNP ID
-
rs144455277
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:162267514 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGCAGATGATTTCACCATTTATTT[C/G]ATAGTCGGCACACTTCTTTTGCAGT
- Phenotype
-
MIM: 606951
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
IFIH1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1990760] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- IFIH1
- Gene Name
- interferon induced with helicase C domain 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_022168.3 |
3264 |
Missense Mutation |
CAA,GAA |
Q955E |
NP_071451.2 |
View Full Product Details