Product Details

SNP ID
rs144546271
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:157258456 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCTGGGAAGGGGCAAGGTTGTGC[C/T]GTTGTGGCATCCTGCACATCTGCAG
Phenotype
MIM: 615129
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GALNT5 PubMed Links

Gene Details

Gene
GALNT5
Gene Name
polypeptide N-acetylgalactosaminyltransferase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017003237.1 909 Missense Mutation CCG,CTG P125L XP_016858726.1
XM_017003238.1 909 Missense Mutation CCG,CTG P125L XP_016858727.1

View Full Product Details