Product Details

SNP ID
rs144639872
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:238102038 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCTCCACGAGGGCCACTCGGCCA[C/T]GCTAGAGACCCGGGAGGGAGCCCGG
Phenotype
MIM: 611056
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ESPNL PubMed Links

Gene Details

Gene
ESPNL
Gene Name
espin-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308370.1 502 Intron NP_001295299.1
NM_194312.3 502 Missense Mutation ACG,ATG T131M NP_919288.2
XM_011511087.1 502 Intron XP_011509389.1
Gene
SCLY
Gene Name
selenocysteine lyase
There are no transcripts associated with this gene.

Gene
UBE2F-SCLY
Gene Name
UBE2F-SCLY readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details