Product Details

SNP ID
rs145240391
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:219209271 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCTCTTTGCTCTAGGCCCAGAGC[C/T]GCAGCTCGAAGCCGTCCAACTGCAG
Phenotype
MIM: 605452 MIM: 613474
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ABCB6 PubMed Links

Gene Details

Gene
ABCB6
Gene Name
ATP binding cassette subfamily B member 6 (Langereis blood group)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005689.2 819 Intron NP_005680.1
Gene
ZFAND2B
Gene Name
zinc finger AN1-type containing 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270998.1 819 Missense Mutation CGC,TGC R247C NP_001257927.1
NM_001270999.1 819 Missense Mutation CGC,TGC R203C NP_001257928.1
NM_138802.2 819 Missense Mutation CGC,TGC R247C NP_620157.1
XM_006712285.1 819 Silent Mutation GCC,GCT A222A XP_006712348.1
XM_006712286.3 819 Silent Mutation GCC,GCT A222A XP_006712349.1
XM_017003368.1 819 Missense Mutation CGC,TGC R247C XP_016858857.1

View Full Product Details