Product Details

SNP ID
rs145641696
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:24791728 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGACTGTGTGATCGTGGATAAGT[C/T]AGGTGACTTCGCTGGCCTCTTCGTC
Phenotype
MIM: 611504
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CENPO PubMed Links

Gene Details

Gene
CENPO
Gene Name
centromere protein O
There are no transcripts associated with this gene.

Gene
PTRHD1
Gene Name
peptidyl-tRNA hydrolase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013663.1 Intron NP_001013685.1

View Full Product Details