Product Details

SNP ID
rs145778937
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:120797452 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTGGCTGCAGCGGCAGCAGCAGC[A/G]GTAGCTGCCCAAGGAGGTACTTTCT
Phenotype
MIM: 165230
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GLI2 PubMed Links

Gene Details

Gene
GLI2
Gene Name
GLI family zinc finger 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005270.4 49 Silent Mutation GCA,GCG A44A NP_005261.2
XM_006712422.2 49 Silent Mutation GCA,GCG A44A XP_006712485.1
XM_011510969.2 49 Intron XP_011509271.2
XM_011510970.2 49 Intron XP_011509272.1
XM_011510971.2 49 Intron XP_011509273.1
XM_011510972.2 49 Intron XP_011509274.2
XM_011510973.2 49 UTR 5 XP_011509275.1
XM_011510974.2 49 UTR 5 XP_011509276.1
XM_017003818.1 49 Intron XP_016859307.1

View Full Product Details