Product Details

SNP ID
rs145867221
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:10913368 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCCTCGGGCCTGCAGACCCTCAC[A/C]TATGCCCTCAAGCGCAGCTTCAAGG
Phenotype
MIM: 603787
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KCNF1 PubMed Links

Gene Details

Gene
KCNF1
Gene Name
potassium voltage-gated channel modifier subfamily F member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002236.4 1432 Silent Mutation ACA,ACC T314T NP_002227.2

View Full Product Details