Product Details

SNP ID
rs146172300
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:44275748 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCCTATGCGGGGATGCCCAAGGA[A/G]GTGCTGTTCCAGTTCTCTGGCCAGG
Phenotype
MIM: 104614
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC3A1 PubMed Links

Gene Details

Gene
SLC3A1
Gene Name
solute carrier family 3 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000341.3 291 Silent Mutation GAA,GAG E71E NP_000332.2
XM_011533047.2 291 Silent Mutation GAA,GAG E71E XP_011531349.1

View Full Product Details