Product Details

SNP ID
rs147745025
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:113022720 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGTTGGTCCGCATGGATGAGAAA[C/T]CTTTGTCCTTCTTCCTGAGATGGTG
Phenotype
MIM: 605508
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL36B PubMed Links

Gene Details

Gene
IL36B
Gene Name
interleukin 36, beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014438.4 557 Missense Mutation GAT,GGT D150G NP_055253.2
NM_173178.2 557 Intron NP_775270.1
XM_011510962.1 557 Intron XP_011509264.1

View Full Product Details