Product Details

SNP ID
rs147772907
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:127869521 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACACCTTTTTCATTAATGAATTC[A/G]ATTCGAATCCCATGGACCCCTACCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AMMECR1L PubMed Links

Gene Details

Gene
AMMECR1L
Gene Name
AMMECR1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199140.1 1289 Silent Mutation ATC,ATT I219I NP_001186069.1
NM_031445.2 1289 Intron NP_113633.2
XM_005263804.4 1289 Silent Mutation ATC,ATT I219I XP_005263861.1
XM_005263806.4 1289 Silent Mutation ATC,ATT I182I XP_005263863.1
XM_005263807.4 1289 Silent Mutation ATC,ATT I182I XP_005263864.1
XM_011511952.2 1289 Silent Mutation ATC,ATT I219I XP_011510254.1
XM_011511954.2 1289 Silent Mutation ATC,ATT I91I XP_011510256.1
XM_011511955.2 1289 Intron XP_011510257.1
XM_011511956.2 1289 Intron XP_011510258.1
XM_017005063.1 1289 UTR 3 XP_016860552.1
Gene
LOC107985803
Gene Name
uncharacterized LOC107985803
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017005446.1 1289 Intron XP_016860935.1

View Full Product Details