Product Details

SNP ID
rs147976791
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27082163 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGGAGCTGCAAGGCCTGCGGGGC[A/G]TCCTGCAAGGACTGAGCGGGCGCCT
Phenotype
MIM: 130660 MIM: 614058
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EMILIN1 PubMed Links

Gene Details

Gene
EMILIN1
Gene Name
elastin microfibril interfacer 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007046.3 1091 Missense Mutation ATC,GTC I198V NP_008977.1
Gene
KHK
Gene Name
ketohexokinase
There are no transcripts associated with this gene.

View Full Product Details