Product Details
- SNP ID
-
rs148413056
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:71133033 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGATAGAAAATTTTGATGATGAGCA[C/G]ATTTGGCAACAACTGGAATTGCAAA
- Phenotype
-
MIM: 608419
MIM: 605503
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MCEE
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs10199088] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MCEE
- Gene Name
- methylmalonyl-CoA epimerase
There are no transcripts associated with this gene.
- Gene
- MPHOSPH10
- Gene Name
- M-phase phosphoprotein 10
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005791.2 |
577 |
Missense Mutation |
CAC,CAG |
H75Q |
NP_005782.1 |
View Full Product Details