Product Details

SNP ID
rs148673256
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:96663252 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCTGAATCTGGCAAACACCTCGG[A/G]TAAGTACACAAAGGACTGGCGCTGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FER1L5 PubMed Links

Gene Details

Gene
FER1L5
Gene Name
fer-1 like family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001293083.1 Intron NP_001280012.1
XM_011512110.2 Intron XP_011510412.1
XM_011512111.2 Intron XP_011510413.1
XM_011512113.2 Intron XP_011510415.1
XM_011512114.2 Intron XP_011510416.1
XM_011512115.2 Intron XP_011510417.1
XM_011512116.2 Intron XP_011510418.1
XM_011512117.2 Intron XP_011510419.1
XM_011512118.2 Intron XP_011510420.1
XM_011512119.2 Intron XP_011510421.1
XM_011512120.2 Intron XP_011510422.1
XM_011512122.2 Intron XP_011510424.1
XM_011512124.2 Intron XP_011510426.1
XM_011512125.2 Intron XP_011510427.1
XM_011512126.2 Intron XP_011510428.1
XM_011512127.2 Intron XP_011510429.1
XM_011512128.2 Intron XP_011510430.1
XM_017005222.1 Intron XP_016860711.1
XM_017005223.1 Intron XP_016860712.1
XM_017005224.1 Intron XP_016860713.1

View Full Product Details